missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl borate, 98+%

Product Code. 10713524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
80 g
750 g
Packaging:
Glass bottle
Unit Size:
750g
80g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10713524 750 g Glass bottle 750g
10236630 80 g Glass bottle 80g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10713524 Supplier Thermo Scientific Acros Supplier No. 188747500
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5419-55-6 | C9H21BO3 | 188.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5419-55-6
Molecular Formula C9H21BO3
Molecular Weight (g/mol) 188.07
MDL Number MFCD00008872
InChI Key NHDIQVFFNDKAQU-UHFFFAOYSA-N
Synonym triisopropyl borate, isopropyl borate, triisopropoxyborane, triisopropoxyboron, boron isopropoxide, triisopropyl orthoborate, boric acid triisopropyl ester, triisopropoxy borane, boron triisopropoxide, boric acid h3bo3 , tris 1-methylethyl ester
PubChem CID 21531
IUPAC Name tripropan-2-yl borate
SMILES B(OC(C)C)(OC(C)C)OC(C)C

Specifications

Melting Point -59°C
Color Colorless
Density 0.8140g/mL
Boiling Point 140°C
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)2CHO]3B
Refractive Index 1.3755 to 1.3775
Quantity 750 g
Beilstein 01, 363
Specific Gravity 0.814
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 188.07
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Triisopropyl borate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.