missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl phosphite, 90+%, Thermo Scientific Chemicals

Product Code. 11462298
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462298 250 mL 250mL
11423910 100 mL 100mL
11433910 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11462298 Supplier Thermo Scientific Alfa Aesar Supplier No. A15247.AK
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-17-6
Molecular Formula C9H21O3P
Molecular Weight (g/mol) 208.238
MDL Number MFCD00008874
InChI Key SJHCUXCOGGKFAI-UHFFFAOYSA-N
Synonym triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite
PubChem CID 8304
IUPAC Name tripropan-2-yl phosphite
SMILES CC(C)OP(OC(C)C)OC(C)C

Specifications

Density 0.908
Boiling Point 63°C to 64°C (11 mmHg)
Flash Point 46°C (114°F)
Odor Characteristic
Refractive Index 1.411
Quantity 250 mL
UN Number UN1993
Beilstein 1701528
Sensitivity Air and moisture sensitive
Formula Weight 208.24
Percent Purity ≥90%
Chemical Name or Material Triisopropyl phosphite
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.