missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilane, 95%, ACROS Organics™

Product Code. 10790214 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
10g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10790214 10g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10790214 Supplier Thermo Scientific Acros Supplier No. 372410100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 6485-79-6
Color Colorless
Density 0.7730g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C9H22Si
Linear Formula [(CH3)2CH]3SiH
Refractive Index 1.4330 to 1.4360 (20°C, 589nm)
MDL Number MFCD00009657
Quantity 10 g
Beilstein 04, III, 1851
Synonym triisopropylsilane, tris propan-2-yl silane, triisopropyl silane, tri propan-2-yl silicon, tri-isopropylsilyl radical, triisopropylsilyl, tri-iso-propylsilane, ambotzrl-1102, pubchem12855
InChI Key YDJXDYKQMRNUSA-UHFFFAOYSA-N
SMILES CC(C)[SiH](C(C)C)C(C)C
IUPAC Name tris(propan-2-yl)silane
Molecular Weight (g/mol) 158.36
PubChem CID 6327611
Formula Weight 158.36
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material Triisopropylsilane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.