missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilane, 98%, AcroSeal™

Product Code. 11946161 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11946161 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11946161 Supplier Thermo Scientific Acros Supplier No. 446171000
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6485-79-6 | C9H22Si | 158.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6485-79-6
Density 0.7730g/mL
Boiling Point 84°C to 86°C (35.0 mmHg)
Flash Point 37°C
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Molecular Formula C9H22Si
Linear Formula [(CH3)2CH]3SiH
Refractive Index 1.4340 to 1.436
MDL Number MFCD00009657
Quantity 100 mL
Specific Gravity 0.773
Synonym triisopropylsilane, tris propan-2-yl silane, triisopropyl silane, tri propan-2-yl silicon, tri-isopropylsilyl radical, triisopropylsilyl, tri-iso-propylsilane, ambotzrl-1102, pubchem12855
Solubility Information Solubility in water: negligible
InChI Key YDJXDYKQMRNUSA-UHFFFAOYSA-N
SMILES CC(C)[SiH](C(C)C)C(C)C
IUPAC Name tris(propan-2-yl)silane
Molecular Weight (g/mol) 158.36
PubChem CID 6327611
Formula Weight 158.36
Percent Purity 98%
Chemical Name or Material Triisopropylsilane, AcroSeal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triisopropylsilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.