missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilane, 98%

Product Code. 11312157
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312157 25 g 25g
11302157 5 g 5g
11322157 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11312157 Supplier Thermo Scientific Alfa Aesar Supplier No. L09585.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6485-79-6 | C9H22Si | 158.36 g/mol

Triisopropylsilane is used as a protecting group in peptide synthesis and is also used as a reducing agent for the selective reduction of anomeric C-phenyl ketals. It is used in the selective silylation of primary hydroxyl groups without affecting the secondary hydroxyl group. It is used in the preparation of anti-1,2-diols catalyzed by a Ni(O) N-heterocyclic carbine complex with high diastereoselectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triisopropylsilane is used as a protecting group in peptide synthesis and is also used as a reducing agent for the selective reduction of anomeric C-phenyl ketals. It is used in the selective silylation of primary hydroxyl groups without affecting the secondary hydroxyl group. It is used in the preparation of anti-1,2-diols catalyzed by a Ni(O) N-heterocyclic carbine complex with high diastereoselectivity.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6485-79-6
Molecular Formula C9H22Si
Molecular Weight (g/mol) 158.36
MDL Number MFCD00009657
InChI Key YDJXDYKQMRNUSA-UHFFFAOYSA-N
Synonym triisopropylsilane, tris propan-2-yl silane, triisopropyl silane, tri propan-2-yl silicon, tri-isopropylsilyl radical, triisopropylsilyl, tri-iso-propylsilane, ambotzrl-1102, pubchem12855
PubChem CID 6327611
SMILES CC(C)[SiH](C(C)C)C(C)C

Specifications

Boiling Point 166°C
Quantity 25 g
Flash Point 35°C (95°F)
Odor Mild
Refractive Index 1.434
Density 0.773
Linear Formula [(CH3)2CH]3SiH
UN Number UN1993
Beilstein 1733718
Solubility Information Immiscible with water.
IUPAC Name tri(propan-2-yl)silicon
Formula Weight 158.36
Percent Purity 98%
Chemical Name or Material Triisopropylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triisopropylsilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Triisopropylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.