missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilane, TRC

Product Code. 30456590
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456590 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456590 Supplier Toronto Research Chemicals Supplier No. TRCT77033050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triisopropylsilane
CAS 6485-79-6
Molecular Formula C9H22Si
Quantity 50 g
Synonym Tris(1-methylethyl)silane, Triisopropylhydrosilane
InChI Formula InChI=1S/C9H22Si/c1-7(2)10(8(3)4)9(5)6/h7-10H,1-6H3
SMILES CC(C)[SiH](C(C)C)C(C)C
IUPAC Name tri(propan-2-yl)silane
Molecular Weight (g/mol) 158.36
Formula Weight 158.1491
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.