missing translation for 'onlineSavingsMsg'
Learn More

(Triisopropylsilyl)acetylene, 97%

Product Code. 10018981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10018981 25 g Glass bottle 25g
10571702 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10018981 Supplier Thermo Scientific Acros Supplier No. 368740250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89343-06-6 | C11H22Si | 182.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89343-06-6
Molecular Formula C11H22Si
Molecular Weight (g/mol) 182.38
MDL Number MFCD00075452
InChI Key KZGWPHUWNWRTEP-UHFFFAOYSA-N
Synonym triisopropylsilyl acetylene, ethynyltriisopropylsilane, triisopropylsilylacetylene, ethynyltris propan-2-yl silane, ethynyl-tri propan-2-yl silane, silane, ethynyltris 1-methylethyl, triisopropylsilylacetylen, acmc-209qzk, ethynyltriisopropyl-silane
PubChem CID 2734682
IUPAC Name ethynyl-tri(propan-2-yl)silane
SMILES CC(C)[Si](C#C)(C(C)C)C(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8130g/mL
Boiling Point 50°C to 52°C (6.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)2CH]3SiC∽CH
Refractive Index 1.4515 to 1.4535
Quantity 25 g
Specific Gravity 0.813
Formula Weight 182.38
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (Triisopropylsilyl)acetylene, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.