missing translation for 'onlineSavingsMsg'
Learn More

Trilaurin, TRC

Product Code. 30456531
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456531 500 mg 500mg
30454761 1 g 1g
30452813 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456531 Supplier Toronto Research Chemicals Supplier No. TRCT795420500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-24-9
Molecular Formula C39 H74 O6
Molecular Weight (g/mol) 639
Synonym Dodecanoic acid 1,1',1' '-(1,2,3-propanetriyl) ester, Dodecanoic acid 1,2,3-propanetriyl ester, tri-Laurin, Dynasan 112, Glycerin tridodecanoate, Glycerin trilaurate, Glycerol trilaurate, Glyceryl laurate, Glyceryl tridodecanoate, Glyceryl trilaurate, LaLaLa triacylglycerol, Lauric acid triglyceride, Lauric acid triglycerin ester, Lauric triglyceride, NSC 4061, Tridodecanoin, Tridodecanoyl glycerol, Triglyceride LaLaLa, Trilaurin, Trilauroylglycerol
IUPAC Name 2,3-di(dodecanoyloxy)propyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C39H74O6/c1-4-7-10-13-16-19-22-25-28-31-37(40)43-34-36(45-39(42)33-30-27-24-21-18-15-12-9-6-3)35-44-38(41)32-29-26-23-20-17-14-11-8-5-2/h36H,4-35H2,1-3H3
Formula Weight 638.5485
Chemical Name or Material Trilaurin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.