missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM15 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
196.00 € - 468.00 €
Specifications
| Antigen | TRIM15 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TRIM15 Polyclonal antibody specifically detects TRIM15 in Human samples. It is validated for Western BlotSpecifications
| TRIM15 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| PBS (pH 7.3), 50% glycerol | |
| 89870 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| RNF93RING finger protein 93, tripartite motif containing 15, tripartite motif-containing 15, Zinc finger protein 178ZNF178, Zinc finger protein B7, ZNFB7tripartite motif-containing protein 15 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-330 of human TRIM15 (NP_150232.2). MPATPSLKVVHELPACTLCAGPLEDAVTIPCGHTFCRLCLPALSQMGAQSSGKILLCPLCQEEEQAETPMAPVPLGPLGETYCEEHGEKIYFFCENDAEFLCVFCREGPTHQAHTVGFLDEAIQPYRDRLRSRLEALSTERDEIEDVKCQEDQKLQVLLTQIESKKHQVETAFERLQQELEQQRCLLLARLRELEQQIWKERDEYITKVSEEVTRLGAQVKELEEKCQQPASELLQDVRVNQSRCEMKTFVSPEAISPDLVKKIRDFHRKILTLPEMMRMFSENLAHHLEIDSGVITLDPQTASRSLVLSEDRKSVRYTRQKKSLPDSPL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title