missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TRIM45 Recombinant Protein

Product Code. p-3722938
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16184883 10 μg 10µg
16194883 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16184883 Supplier Abnova™ Supplier No. H00080263P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH34943
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 80263
Molecular Weight (g/mol) 87.56
Name TRIM45 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MSENRKPLLGFVSKLTSGTALGNSGKTHCPLCLGLFKAPRLLPCLHTVCTTCLEQLEPFSVVDIRGGDSDTSSEGSIFQELKPRSLQSQIGILCPVCDAQVDLPMGGVKALTIDHLAVNDVMLESLRGEGQGLVCDLCNDREVEKRCQTCKANLCHFCCQAHRRQKKTTYHTMVDLKDLKGYSRIGKPILCPVHPAEELRLFCEFCDRPVCQDCVVGEHREHPCDFTSNVIHKHGDSVWELLKGTQPHVEALEEALAQIHIINSALQKRVEAVAADVRTFSEGYIKAIEEHRDKLLKQLEDIRAQKENSLQLQKAQLEQLLADMRTGVEFTEHLLTSGSDLEILITKRVVVERLRKLNKVQYSTRPGVNDKIRFCPQEKAGQCRGYEIYDLHRAREKQTASFTLLCKDAAGEIMGRGGDNVQVAVVPKDKKDSPVRTMVQDNKDGTYYISYTPKEPGVYTVWVCIKEQHVQGSPFTVTVRRKHRPHSGVFHCCTFCSSGGQKTARCACGGTMPGGYLGCGHGHKGHPGHPHWSCCGKFNEKSECTWTGGQSAPRSLLRTVAL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias FLJ13181/RNF99
Common Name TRIM45
Gene Symbol TRIM45
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.