missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Trimethoprim, 98%

Product Code. p-7077531 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15345602 5 g 5g
15780889 25 g 25g
15790889 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15345602 Supplier Thermo Scientific Chemicals Supplier No. 455120050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trimethoprim, CAS # 738-70-5, is a trimethoxybenzyl-pyrimidine used as an antimicrobial. | CAS: 738-70-5 | C14H18N4O3 | 290.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Trimethoprim is a synthetic pyrimidine analog that binds dihydrofolate-reductase and inhibits the synthesis of folic acid; when paired with sulfamethoxazole, its activity is enhanced • Trimethoprim has antibacterial and antiprotozoal properties

Applications

• Trimethoprim inhibits the conversion of para-aminobenzoic acid (PABA) to folic acid near the end of the synthesis pathway and can be used to study related effects

• Trimethoprim has been radiolabeled and used to trace routes of infection, and it also has been used experimentally as a fluorescent labeler to quantify dihydrofolate-reductase expression

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 738-70-5
Molecular Formula C14H18N4O3
Molecular Weight (g/mol) 290.32
InChI Key IEDVJHCEMCRBQM-UHFFFAOYSA-N
Synonym trimethoprim, proloprim, trimpex, bactramin, monotrim, monotrimin, trimetoprim, trimopan, monoprim, syraprim
PubChem CID 5578
ChEBI CHEBI:45924
IUPAC Name 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
SMILES COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N

Specifications

Melting Point 234.0°C to 236.0°C
Loss on Drying 0.5% max. (105°C, 4 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: insoluble.
Formula Weight 290.32
Percent Purity 98%
Chemical Name or Material Trimethoprim

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.