missing translation for 'onlineSavingsMsg'
Learn More

Trimethoprim, TRC

Product Code. 30007664
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007664 5 g 5g
17921675 1 g 1g
17924025 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007664 Supplier Toronto Research Chemicals Supplier No. TRCT7956155G
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 738-70-5
Molecular Formula C14 H18 N4 O3
Molecular Weight (g/mol) 290.32
Synonym Trimethoprim, 5-(3,4,5-Trimethoxybenzyl)pyrimidine-2,4-diamine, Pyrimidine, 2,4-diamino-5-(3,4,5-trimethoxybenzyl)- (7CI,8CI), 2,4-Pyrimidinediamine, 5-[(3,4,5-trimethoxyphenyl)methyl]-, 2,4-Diamino-5-(3',4',5'-trimethoxybenzyl)pyrimidine, 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine, BW 56-72, Instalac, Monotrim, NIH 204, NSC 106568, Proloprim, Syraprim, Tiempe, Trimanyl, Trimethioprim, Trimetoprim, Trimogal, Trimopan, Trimpex, Triprim, Uretrim, Wellcoprim
IUPAC Name 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
SMILES COc1cc(Cc2cnc(N)nc2N)cc(OC)c1OC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H18N4O3/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15/h5-7H,4H2,1-3H3,(H4,15,16,17,18)
Formula Weight 290.1379
Percent Purity >95%
Chemical Name or Material Trimethoprim
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.