missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl borate, 99%, AcroSeal™

Product Code. 10333295 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10333295 800 mL AcroSeal™ Glass Bottle 800mL
10566695 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10333295 Supplier Thermo Scientific Supplier No. 346128000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121-43-7 | C3H9O3B | 103.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-43-7
Molecular Formula C3H9O3B
Molecular Weight (g/mol) 103.91
MDL Number MFCD00008346
InChI Key WRECIMRULFAWHA-UHFFFAOYSA-N
Synonym trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
PubChem CID 8470
ChEBI CHEBI:38913
IUPAC Name trimethyl borate
SMILES B(OC)(OC)OC

Specifications

Melting Point -34.0°C
Density 0.9150g/mL
Boiling Point 68.0°C to 69.0°C
Flash Point −8°C
Assay Percent Range 99%
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3O)3B
Refractive Index 1.3562 to 1.3582
Quantity 800 mL
Beilstein 01, 287
Fieser 01,673; 13,218
Merck Index 15, 9882
Specific Gravity 0.915
Solubility Information Solubility in water: reacts. Other solubilities: miscible with thf,diethyl ether,hexane,methanol,,isopropylamine,and other organic liquids
Viscosity 0.36 mPa.s (25°C)
Formula Weight 103.91
Percent Purity 99%
Chemical Name or Material Trimethyl borate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.