missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl citrate

Product Code. 15711809 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15711809

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1587-20-8 | C9H14O7 | 234.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1587-20-8
Molecular Formula C9H14O7
Molecular Weight (g/mol) 234.2
InChI Key HDDLVZWGOPWKFW-UHFFFAOYSA-N
PubChem CID 74112

Specifications

Melting Point 75°C to 78°C
Boiling Point 176°C (16.0 mmHg)
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in chloroform,methanol
Formula Weight 234.2
Chemical Name or Material Trimethyl citrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.