missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl-d9 Phosphate, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Trimethyl Phosphate-d9 |
| CAS | 32176-12-8 |
| Molecular Formula | C3 D9 O4 P |
| Quantity | 0.5 g |
| Synonym | NSC 58985-d9, TMP-d9, TMPA-d9, TMPO-d9, Trimethoxyphosphine Oxide-d9, Trimethyl Orthophosphate-d9, Trimethyl Phosphate-d9 |
| InChI Formula | InChI=1 S/C3H9O4P/c1-5-8(4,6-2)7-3/h1-3H3/i1D3,2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])OP(=O)(OC([2 H])([2 H])[2 H])OC([2 H])([2 H])[2 H] |
| IUPAC Name | tris(trideuteriomethyl) phosphate |
| Molecular Weight (g/mol) | 149.13 |
| Formula Weight | 149.0803 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction