missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl orthoformate, 99+%, anhydrous, AcroSeal™

Product Code. 11984941 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11984941 100 mL 100mL
11994941 800 mL 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11984941 Supplier Thermo Scientific Acros Supplier No. 429281000
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 149-73-5 | C4H10O3 | 106.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-73-5
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
InChI Key PYOKUURKVVELLB-UHFFFAOYSA-N
Synonym trimethyl orthoformate, methyl orthoformate, methane, trimethoxy, orthoformic acid, trimethyl ester, orthoformic acid methyl ester, orthomravencan methylnaty, methoxymethylal, methylester kyseliny orthomravenci, unii-xam28819yj, orthoformic acid trimethyl ester
PubChem CID 9005
IUPAC Name trimethoxymethane
SMILES COC(OC)OC

Specifications

Melting Point -53.0°C
Density 0.9700g/mL
Boiling Point 101.0°C to 102.0°C
Flash Point 13°C
Assay Percent Range 99% min. (GC)
Packaging AcroSeal glass Bottle
Linear Formula CH(OCH3)3
Refractive Index 1.3780 to 1.3800
Quantity 100 mL
Merck Index 15,698
Specific Gravity 0.97
Solubility Information Solubility in water: 10g/L (hydrolysis). Other solubilities: soluble in organic solvents
Formula Weight 106.12
Percent Purity 99+%
Chemical Name or Material Trimethyl orthoformate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethyl orthoformate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.