missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl orthoformate, 99%

Product Code. 10572541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10572541 1 kg Glass bottle 1kg
10356740 100 g Glass bottle 100g
10644812 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10572541 Supplier Thermo Scientific Acros Supplier No. 148660010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 149-73-5 | C4H10O3 | 106.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-73-5
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00008483
InChI Key PYOKUURKVVELLB-UHFFFAOYSA-N
Synonym trimethyl orthoformate, methyl orthoformate, methane, trimethoxy, orthoformic acid, trimethyl ester, orthoformic acid methyl ester, orthomravencan methylnaty, methoxymethylal, methylester kyseliny orthomravenci, unii-xam28819yj, orthoformic acid trimethyl ester
PubChem CID 9005
IUPAC Name trimethoxymethane
SMILES COC(OC)OC

Specifications

Melting Point -53.0°C
Color Colorless
Density 0.9700g/mL
Boiling Point 101.0°C to 102.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH(OCH3)3
Refractive Index 1.3780 to 1.3800
Quantity 1 kg
Beilstein 02, 19
Fieser 03,313; 04,540; 05,714; 08,507; 10,425; 14,330; 15,342
Merck Index 15, 6980
Specific Gravity 0.97
Solubility Information Solubility in water: 10g/L (hydrolysis). Other solubilities: soluble in organic solvents
Formula Weight 106.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Trimethyl orthoformate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethyl orthoformate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.