missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl orthopropionate, 97%

Product Code. 10751894 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10751894 100 mL Glass bottle 100mL
10242603 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10751894 Supplier Thermo Scientific Acros Supplier No. 384621000
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 24823-81-2 | C6H14O3 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24823-81-2
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00043305
InChI Key ZGMNAIODRDOMEK-UHFFFAOYSA-N
PubChem CID 141145
IUPAC Name 1,1,1-trimethoxypropane
SMILES CCC(OC)(OC)OC

Specifications

Color Colorless
Density 0.9440g/mL
Boiling Point 121°C to 122°C
Flash Point 19°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2C(OCH3)3
Refractive Index 1.3970 to 1.399
Quantity 100 mL
Specific Gravity 0.944
Synonym trimethyl orthopropionate, propane, 1,1,1-trimethoxy, trimethylorthopropionate, trimethoxypropane, acmc-209gew, trimethyl orthopropanoate, propane,1,1,1-trimethoxy, ksc489s6b, zgmnaiodrdomek-uhfffaoysa, orthopropionic acid trimethyl ester
Formula Weight 134.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Trimethyl orthopropionate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethyl orthopropionate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.