missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Orthopropionate, TRC

Product Code. 30456985
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456985 250 mg 250mg
30453270 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456985 Supplier Toronto Research Chemicals Supplier No. TRCT796775250MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethyl Orthopropionate
CAS 24823-81-2
Molecular Formula C6H14O3
Quantity 250 mg
Synonym 1,1,1-Trimethoxypropane
InChI Formula InChI=1S/C6H14O3/c1-5-6(7-2,8-3)9-4/h5H2,1-4H3
SMILES CCC(OC)(OC)OC
IUPAC Name 1,1,1-trimethoxypropane
Molecular Weight (g/mol) 134.17
Formula Weight 134.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.