missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl phosphonoacetate, 98%

Product Code. 11925801 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11925801 500 g Glass bottle 500g
10593301 25 g Glass bottle 25g
10163380 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11925801 Supplier Thermo Scientific Acros Supplier No. 140235000
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5927-18-4 | C5H11O5P | 182.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5927-18-4
Molecular Formula C5H11O5P
Molecular Weight (g/mol) 182.11
InChI Key SIGOIUCRXKUEIG-UHFFFAOYSA-N
Synonym trimethyl phosphonoacetate, methyl 2-dimethoxyphosphoryl acetate, trimethyl phosphonacetate, trimethylacetyl phosphate, acetic acid, dimethoxyphosphinyl-, methyl ester, methyl-dimethoxyphosphoryl acetat, trimethyl phosphoroacetate, phosphonoacetic acid trimethyl ester, methyl dimethylphosphonoacetate, methyl dimethoxyphosphoryl acetate
PubChem CID 80029
IUPAC Name methyl 2-dimethoxyphosphorylacetate
SMILES COC(=O)CP(=O)(OC)OC

Specifications

Color Colorless to Yellow
Density 1.2600g/mL
Boiling Point 265.0°C to 268.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2P(O)CH2CO2CH3
Refractive Index 1.4360 to 1.4380
Quantity 500 g
Fieser 02,442; 07,394; 01,314; 02,169; 01,1236; 02,171; 13,15
Specific Gravity 1.26
Formula Weight 182.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Trimethyl phosphonoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.