missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Thiophosphate, TRC

Product Code. 30456770
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mg
missing translation for 'unitSize'
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456770 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30456770 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT797605500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O,O,O-Trimethylthiophosphate
CAS 152-18-1
Molecular Formula C3 H9 O3 P S
Quantity 500 mg
Synonym O,O,O-Trimethylphosphorthioate, Trimethoxy(thioxo)-lambda5-phosphane, Phosphorothioic acid','O,O,O-trimethyl ester (8CI','9CI','ACI), 7900-HC, HC 7900, Methyl phosphorothioate ((MeO)3PS), O,O,O-Trimethyl phosphorothioate, O,O,O-Trimethyl phosphorothionate, O,O,O-Trimethyl thiophosphate, SD 4741, Trimethoxyphosphine sulfide, Trimethyl thionophosphate
InChI Formula InChI=1S/C3H9O3PS/c1-4-7(8,5-2)6-3/h1-3H3
SMILES COP(=S)(OC)OC
IUPAC Name trimethoxy(sulfanylidene)-λ^{5}-phosphane
Molecular Weight (g/mol) 156.14
Formula Weight 156.001
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.