missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetic acid, 99%

Product Code. 10183280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10183280 5 g Glass bottle 5g
10254760 100 g Glass bottle 100g
10366830 500 g Glass bottle 500g
10463585 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10183280 Supplier Thermo Scientific Acros Supplier No. 140020050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-98-9 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-98-9
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00004194
InChI Key IUGYQRQAERSCNH-UHFFFAOYSA-N
Synonym pivalic acid, trimethylacetic acid, neopentanoic acid, 2,2-dimethylpropionic acid, tert-pentanoic acid, propanoic acid, 2,2-dimethyl, versatic 5, kyselina pivalova, acetic acid, trimethyl, alpha,alpha-dimethylpropionic acid
PubChem CID 6417
ChEBI CHEBI:45133
IUPAC Name 2,2-dimethylpropanoic acid
SMILES CC(C)(C)C(=O)O

Specifications

Melting Point 33.0°C to 35.0°C
Color Tan to Yellow
Boiling Point 163.0°C to 164.0°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCO2H
Quantity 5 g
Beilstein 02, 319
Fieser 01,1229
Merck Index 15, 7625
Solubility Information Solubility in water: 25g/L (20°C). Other solubilities: soluble in alcohol and ether
Viscosity 3.6 mm2/s (60°C)
Formula Weight 102.13
Percent Purity 99%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Trimethylacetic acid, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethylacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.