missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine hydrochloride, 98%

Product Code. 10407530 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10407530 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10407530 Supplier Thermo Scientific Acros Supplier No. 140071000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 593-81-7 | C3H10ClN | 95.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 593-81-7
Melting Point 283.0°C to 284.0°C
Color Cream to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C3H10ClN
Linear Formula (CH3)3N·HCl
MDL Number MFCD00012478
Quantity 100 g
Beilstein 04, 43
Merck Index 15, 9880
Synonym trimethylamine hydrochloride, trimethylammonium chloride, trimethylamine hcl, n,n-dimethylmethanamine hydrochloride, trimethylamine monohydrochloride, methanamine, n,n-dimethyl-, hydrochloride, trimethylaminehydrochloride, unii-uw38srm77k, uw38srm77k, trimethyl ammonium chloride
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol,moderately soluble in chloroform,insoluble in ether
InChI Key SZYJELPVAFJOGJ-UHFFFAOYSA-N
SMILES [H+].[Cl-].CN(C)C
IUPAC Name N,N-dimethylmethanamine;hydrochloride
Molecular Weight (g/mol) 95.57
PubChem CID 10313079
ChEBI CHEBI:64700
Formula Weight 95.57
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Trimethylamine hydrochloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Trimethylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.