missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine N-oxide dihydrate, 98%

Product Code. 10484891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10484891 25 g Glass bottle 25g
10494891 100 g Glass bottle 100g
11995701 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10484891 Supplier Thermo Scientific Acros Supplier No. 421890250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 62637-93-8 | C3H13NO3 | 111.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62637-93-8
Molecular Formula C3H13NO3
Molecular Weight (g/mol) 111.14
MDL Number MFCD00149077
InChI Key PGFPZGKEDZGJQZ-UHFFFAOYSA-N
Synonym trimethylamine-n-oxide dihydrate, trimethylamine n-oxide dihydrate, trimethylamine oxide dihydrate, trimethylamine oxide, dihydrate, n,n-dimethylmethanamine oxide, dihydrate, unii-c73wz0186w, trimethylammoniumoxid hydrat czech, methanamine oxide, n,n-dimethyl, dihydrate, methanamine, n,n-dimethyl-, n-oxide, dihydrate, n,n-dimethylmethanamine oxide dihydrate
PubChem CID 198430
SMILES O.O.C[N+](C)(C)[O-]

Specifications

Melting Point 94.0°C to 98.0°C
Color White
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (Total base)
Packaging Glass bottle
Linear Formula (CH3)3NO·2H2O
Quantity 25 g
Fieser 01,1230; 02,434; 03,309; 06,624; 07,392; 08,507; 09,488; 14,329
Merck Index 15,9881
IUPAC Name N,N-dimethylmethanamine oxide dihydrate
Formula Weight 111.14
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Trimethylamine N-oxide dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Trimethylamine N-oxide dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.