missing translation for 'onlineSavingsMsg'
Learn More

Trimethylboroxin, TRC

Product Code. 30452409
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452409 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452409 Supplier Toronto Research Chemicals Supplier No. TRCT796145250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethylboroxin
CAS 823-96-1
Molecular Formula C3H9B3O3
Quantity 250 mg
Synonym 2,4,6-Trimethyl-boroxin, Trimethyl-boroxin, 2,4,6-Trimethyl-1,3,5,2,4,6-trioxatriborinane, 2,4,6-Trimethylboroxine, Methaneboronic anhydride
InChI Formula InChI=1S/C3H9B3O3/c1-4-7-5(2)9-6(3)8-4/h1-3H3
SMILES CB(OB(C)O1)OB1C
IUPAC Name 2,4,6-trimethyl-1,3,5,2,4,6-trioxatriborinane
Molecular Weight (g/mol) 125.53
Formula Weight 126.0831
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.