missing translation for 'onlineSavingsMsg'
Learn More

Trimethylboroxine, 3.5M (50 wt.%) solution in THF

Product Code. 10125510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10125510 25 mL Glass bottle 25mL
10379273 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10125510 Supplier Thermo Scientific Acros Supplier No. 297520250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 823-96-1 | C3H9B3O3 | 125.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 823-96-1 , 109-99-9
Molecular Formula C3H9B3O3
Molecular Weight (g/mol) 125.53
MDL Number MFCD00013354
InChI Key GBBSAMQTQCPOBF-UHFFFAOYSA-N
Synonym trimethylboroxine, 2,4,6-trimethylboroxin, trimethylboroxin, boroxin, trimethyl, trimethyl-1,3,5,2,4,6-trioxatriborinane, trimethyboroxine, trimethyl boroxin, trimethylboroxaine, trimethyl boroxine, trimethyl-boroxine
PubChem CID 574072
IUPAC Name 2,4,6-trimethyl-1,3,5,2,4,6-trioxatriborinane
SMILES CB1OB(C)OB(C)O1

Specifications

Melting Point -38.0°C
Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 78.0°C to 80.0°C
Flash Point −9°C
Packaging Glass bottle
Quantity 25 mL
Specific Gravity 0.89
Formula Weight 125.54
Percent Purity 40.0 to 60.0% (ex Boron)
Physical Form Solution
Chemical Name or Material Trimethylboroxine, 50 wt. % Solution in THF
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trimethylboroxine, 50 wt% solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tetrahydrofuran, Trimethylboroxine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.