missing translation for 'onlineSavingsMsg'
Learn More

Trimethylgermanium chloride

Product Code. 11397788
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397788 1 g 1g
11317798 5 g 5g
11307798 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397788 Supplier Thermo Scientific Alfa Aesar Supplier No. 034129.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1529-47-1 | C3H9ClGe | 153.185 g/mol

Trimethylgermanium chloride is used to prepare trimethyl(2-thienyl)germane. It is also used as alkylating and precursor to prepare Grignard reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1529-47-1
Molecular Formula C3H9ClGe
Molecular Weight (g/mol) 153.185
MDL Number MFCD00000462
InChI Key ZZBNZZCHSNOXOH-UHFFFAOYSA-N
Synonym trimethylchlorogermane, trimethylgermanium chloride, chloro trimethyl germane, chlorotrimethylgermanium, trimethylgermyl chloride, ch3 3gecl, acmc-209d8g, chlorotrimethylgermane
PubChem CID 73723
IUPAC Name chloro(trimethyl)germane
SMILES C[Ge](C)(C)Cl

Specifications

Melting Point -14°C
Density 1.249 g/mL
Boiling Point 102°C
Flash Point 1°C (35°F)
Refractive Index 1.4337
Quantity 1 g
UN Number UN2924
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.Miscible with ether,tetrahydrofuran,hexane,benzene,dichloromethane and chloroform.
Formula Weight 153.15
Physical Form Liquid
Chemical Name or Material Trimethylgermanium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.