missing translation for 'onlineSavingsMsg'
Learn More

Trimethylphenylammonium Tribromide, TRC

Product Code. 30456996
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456996 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456996 Supplier Toronto Research Chemicals Supplier No. TRCT79614025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Trimethylphenylammonium Tribromide
CAS 4207-56-1
Molecular Formula C9H14Br3N
Quantity 25 g
Synonym N,N,N-Trimethylbenzenaminium (Tribromide) (1:1), Trimethylphenylammonium (Tribromide), N,N,N-Trimethylbenzenaminium (Tribromide), N,N,N-Trimethylbenzenaminium Bromide (Br31-), Jacques Reagent, NSC 138640, NSC 173340, NSC 87897, Phenyltrimethylammonium Perbromide, Phenyltrimethylammonium Tribromide, Trimethylphenylammonium Perbromide
InChI Formula InChI=1S/C9H14N.Br3/c1-10(2,3)9-7-5-4-6-8-9;1-3-2/h4-8H,1-3H3;/q+1;-1
SMILES C[N+](C)(C)c1ccccc1.Br[Br-]Br
Molecular Weight (g/mol) 375.93
Formula Weight 372.8676
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.