missing translation for 'onlineSavingsMsg'
Learn More

Trimethylphenylammonium Tribromide, TRC

Product Code. 30455811
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455811 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455811 Supplier Toronto Research Chemicals Supplier No. TRCT7961402.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Trimethylphenylammonium Tribromide
CAS 4207-56-1
Molecular Formula C9H14Br3N
Quantity 2.5 g
Synonym N,N,N-Trimethylbenzenaminium (Tribromide) (1:1), Trimethylphenylammonium (Tribromide), N,N,N-Trimethylbenzenaminium (Tribromide), N,N,N-Trimethylbenzenaminium Bromide (Br31-), Jacques Reagent, NSC 138640, NSC 173340, NSC 87897, Phenyltrimethylammonium Perbromide, Phenyltrimethylammonium Tribromide, Trimethylphenylammonium Perbromide
InChI Formula InChI=1S/C9H14N.Br3/c1-10(2,3)9-7-5-4-6-8-9;1-3-2/h4-8H,1-3H3;/q+1;-1
SMILES C[N+](C)(C)c1ccccc1.Br[Br-]Br
Molecular Weight (g/mol) 375.93
Formula Weight 372.8676
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.