missing translation for 'onlineSavingsMsg'
Learn More

Trimethylphenylammonium Tribromide, TRC

Product Code. 30455407
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455407 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455407 Supplier Toronto Research Chemicals Supplier No. TRCT79614010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethylphenylammonium Tribromide
CAS 4207-56-1
Molecular Formula C9H14Br3N
Quantity 10 g
Synonym N,N,N-Trimethylbenzenaminium (Tribromide) (1:1), Trimethylphenylammonium (Tribromide), N,N,N-Trimethylbenzenaminium (Tribromide), N,N,N-Trimethylbenzenaminium Bromide (Br31-), Jacques Reagent, NSC 138640, NSC 173340, NSC 87897, Phenyltrimethylammonium Perbromide, Phenyltrimethylammonium Tribromide, Trimethylphenylammonium Perbromide
InChI Formula InChI=1S/C9H14N.Br3/c1-10(2,3)9-7-5-4-6-8-9;1-3-2/h4-8H,1-3H3;/q+1;-1
SMILES C[N+](C)(C)c1ccccc1.Br[Br-]Br
Molecular Weight (g/mol) 375.93
Formula Weight 372.8676
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.