missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl acetate 97%, Thermo Scientific™

Product Code. 10455285 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10455285 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10455285 Supplier Thermo Scientific Acros Supplier No. 391981000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2754-27-0
Density 0.8840g/mL
Boiling Point 102.0°C to 104.0°C
Flash Point 19°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C5H12O2Si
Linear Formula CH3COOSi(CH3)3
Refractive Index 1.3870 to 1.3890
MDL Number MFCD00008695
Quantity 100g
Specific Gravity 0.884
Synonym acetoxytrimethylsilane, trimethylacetoxysilane, acetato trimethylsilane, acetic acid trimethylsilyl ester, silanol, trimethyl-, acetate, silanol, 1,1,1-trimethyl-, 1-acetate, tmsoac, o-trimethylsilylacetate, trimethylsilyl acetate
InChI Key QHUNJMXHQHHWQP-UHFFFAOYSA-N
SMILES CC(=O)O[Si](C)(C)C
IUPAC Name trimethylsilyl acetate
Molecular Weight (g/mol) 132.234
PubChem CID 75988
Formula Weight 132.23
Percent Purity 97%
Chemical Name or Material Trimethylsilyl acetate
Show More Show Less
compliance-icons
Product Identifier
  • Trimethylsilyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.