missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl azide, 94%

Product Code. 11362685
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362685 5 g 5g
11372685 25 g 25g
11382685 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362685 Supplier Thermo Scientific Alfa Aesar Supplier No. L00173.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4648-54-8 | C3H9N3Si | 115.21 g/mol

Trimethylsilyl azide is used as an azidonation reagent for amines, amides, aldehydes and ketones. It is involved in the preparation of alfa- and beta-siloxy azides from carbonyl compounds and epoxides. It is also used in the preparation of heterocyclic compounds and also acts as an effective substitute for hydrazoic acid. Further, it is used in the Oseltamivir total synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl azide is used as an azidonation reagent for amines, amides, aldehydes and ketones. It is involved in the preparation of alfa- and beta-siloxy azides from carbonyl compounds and epoxides. It is also used in the preparation of heterocyclic compounds and also acts as an effective substitute for hydrazoic acid. Further, it is used in the Oseltamivir total synthesis.

Solubility
Miscible with toluene, dichloromethane, diethyl ether and most organic solvents.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4648-54-8
Molecular Formula C3H9N3Si
Molecular Weight (g/mol) 115.21
MDL Number MFCD00001986
InChI Key SEDZOYHHAIAQIW-UHFFFAOYSA-N
Synonym trimethylsilyl azide, silane, azidotrimethyl, trimethylsilylazide, azido trimethyl silane, ccris 8048, tms azide, tms-azide, trimethyl silylazide, trimethylsilyl-azide, azido trimethylsilane
PubChem CID 78378
SMILES C[Si](C)(C)N=[N+]=[N-]

Specifications

Melting Point -95°C
Density 0.872
Boiling Point 95°C to 99°C
Flash Point 6°C (42°F)
Refractive Index 1.414
Quantity 5 g
UN Number UN1992
Beilstein 1903730
Sensitivity Moisture sensitive
Solubility Information Miscible with toluene,dichloromethane,diethyl ether and most organic solvents.
IUPAC Name azidotrimethylsilane
Formula Weight 115.21
Percent Purity 94%
Chemical Name or Material Trimethylsilyl azide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trimethylsilyl azide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Trimethylsilyl azide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.