missing translation for 'onlineSavingsMsg'
Learn More

(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes

Product Code. 10401923 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10401923 25 mL Glass bottle 25mL
10413962 5 mL Glass bottle 5mL
10609433 100 mL Glass bottle 100mL
11925861 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10401923 Supplier Thermo Scientific Acros Supplier No. 385330250
  • ONLINE EXCLUSIVE PRICE
    267.00 €
    249.00 € / 25mL
    Save 18.00 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18107-18-1 | C4H10N2Si | 114.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18107-18-1 , 92112-69-1
Molecular Formula C4H10N2Si
Molecular Weight (g/mol) 114.22
InChI Key ONDSBJMLAHVLMI-UHFFFAOYSA-N
Synonym trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4
PubChem CID 167693
IUPAC Name diazomethyl(trimethyl)silane
SMILES C[Si](C)(C)C=[N+]=[N-]

Specifications

CAS Min % 32.15, 64.47
CAS Max % 35.54, 67.85
Color Yellow
Density 0.6750g/mL
Boiling Point 96.0°C
Flash Point −23°C
Packaging Glass bottle
Linear Formula (CH3)3SiCHN2
Quantity 25 mL
Specific Gravity 0.675
Solubility Information Solubility in water: insoluble
Formula Weight 114.22
Grade Technical
Physical Form Solution
Chemical Name or Material (Trimethylsilyl)diazomethane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (Trimethylsilyl)diazomethane (2.0M solution in hexanes)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H370-Causes damage to organs.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.