missing translation for 'onlineSavingsMsg'
Learn More

(Trimethylsilyl)diazomethane, approx. 2M solution in diethyl ether, AcroSeal™

Product Code. 10801681 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10801681 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10801681 Supplier Thermo Scientific Acros Supplier No. 429201000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 18107-18-1 | C4H10N2Si | 114.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 18107-18-1, 60-29-7
CAS Min % 28.19, 67.36
CAS Max % 32.64, 71.81
Color Colorless
Density 0.7700g/mL
Flash Point −35°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C4H10N2Si
MDL Number MFCD00053946
Quantity 100 mL
Specific Gravity 0.77
Synonym trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4
InChI Key ONDSBJMLAHVLMI-UHFFFAOYSA-N
SMILES C[Si](C)(C)C=[N+]=[N-]
IUPAC Name diazomethyl(trimethyl)silane
Molecular Weight (g/mol) 114.23
PubChem CID 167693
Formula Weight 114.23
Physical Form Liquid
Chemical Name or Material (Trimethylsilyl)diazomethane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (Trimethylsilyl)diazomethane, ca. 2M solution in diethylether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H350-May cause cancer.
  • H330-Fatal if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.