missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl trifluoroacetate, 96%, Thermo Scientific™

Product Code. p-208136 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10667012 25 g Glass bottle 25g
15274684 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10667012 Supplier Thermo Scientific Acros Supplier No. 215970250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 400-53-3
Molecular Formula C5H9F3O2Si
Molecular Weight (g/mol) 186.21
MDL Number MFCD00000413
InChI Key VIYXXANHGYSBLY-UHFFFAOYSA-N
Synonym trimethylsilyl trifluoroacetate, trimethylsilyltrifluoroacetate, acetic acid, trifluoro-, trimethylsilyl ester, acetic acid, 2,2,2-trifluoro-, trimethylsilyl ester, trifluoroacetic acid trimethylsilyl ester, tms trifluoroacetate, acmc-1ago5, cf3c o osi ch3 3, trimethylsilyl-trifluoroacetate, trifluoroacetic acid, tms derivative
PubChem CID 67863
IUPAC Name trimethylsilyl 2,2,2-trifluoroacetate
SMILES C[Si](C)(C)OC(=O)C(F)(F)F

Specifications

Color White
Density 1.0800g/mL
Boiling Point 88.0°C to 90.0°C
Flash Point 0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CF3CO2Si(CH3)3
Refractive Index 1.3350 to 1.3380
Quantity 25 g
Beilstein 04,IV,4004
Specific Gravity 1.08
Solubility Information Solubility in water: reacts
Formula Weight 186.21
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Trimethylsilyl trifluoroacetate, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethylsilyl trifluoroacetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.