missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilylacetylene, 98%

Product Code. 10421251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10421251 25 g Glass bottle 25g
10123790 5 g Glass bottle 5g
10206133 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10421251 Supplier Thermo Scientific Acros Supplier No. 203570250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1066-54-2 | C5H10Si | 98.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-54-2
Molecular Formula C5H10Si
Molecular Weight (g/mol) 98.22
MDL Number MFCD00008569
InChI Key CWMFRHBXRUITQE-UHFFFAOYSA-N
Synonym trimethylsilylacetylene, trimethylsilyl acetylene, silane, ethynyltrimethyl, ethynyl-trimethyl-silane, ethynyl trimethyl silane, tms acetylene, ethynyltrimethyl silane, trimethylsilyl-acetylene, tmsacetylene
PubChem CID 66111
IUPAC Name ethynyl(trimethyl)silane
SMILES C[Si](C)(C)C#C

Specifications

Color Colorless
Density 0.6900g/mL
Boiling Point 53°C
Flash Point -34°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3SiC∽CH
Refractive Index 1.3873 to 1.3903
Quantity 25 g
Specific Gravity 0.69
Solubility Information Solubility in water: reacts. Other solubilities: soluble in most common organic solvents
Formula Weight 98.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Trimethylsilylacetylene
Show More Show Less
compliance-icons
Product Identifier
  • Trimethylsilylacetylene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.