missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfonium Iodide, 98%

Product Code. 10214860 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10214860 100 g Plastic bottle 100g
10133430 25 g Plastic bottle 25g
10435755 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10214860 Supplier Thermo Scientific Acros Supplier No. 140251000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2181-42-2 | C3H9IS | 204.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2181-42-2
Molecular Formula C3H9IS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00011632
InChI Key VFJYIHQDILEQNR-UHFFFAOYSA-M
Synonym trimethylsulfonium iodide, trimethylsulphonium iodide, sulfonium, trimethyl-, iodide, trimethylsulfoniumiodide, trimethylsulfanium iodide, trimethyl-sulfonium iodide, unii-4z1liz2lun, 4z1liz2lun, sulfonium, trimethyl-, iodide 1:1, chembl25340
PubChem CID 75127
IUPAC Name trimethylsulfanium;iodide
SMILES C[S+](C)C.[I-]

Specifications

Melting Point 215.0°C to 220.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula (CH3)3SI
Quantity 100 g
Fieser 01,314; 02,169; 01,1236; 02,171; 13,15
Solubility Information Solubility in water: soluble
Formula Weight 204.07
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Trimethylsulfonium iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.