missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfonium iodide, 98%

Product Code. 11440483
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440483 50 g 50g
11450483 250 g 250g
11430483 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440483 Supplier Thermo Scientific Alfa Aesar Supplier No. A12639.18
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2181-42-2 | C3H9IS | 204.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2181-42-2
Molecular Formula C3H9IS
Molecular Weight (g/mol) 204.069
MDL Number MFCD00011632
InChI Key VFJYIHQDILEQNR-UHFFFAOYSA-M
Synonym trimethylsulfonium iodide, trimethylsulphonium iodide, sulfonium, trimethyl-, iodide, trimethylsulfoniumiodide, trimethylsulfanium iodide, trimethyl-sulfonium iodide, unii-4z1liz2lun, 4z1liz2lun, sulfonium, trimethyl-, iodide 1:1, chembl25340
PubChem CID 75127
IUPAC Name trimethylsulfanium;iodide
SMILES C[S+](C)C.[I-]

Specifications

Melting Point ∼215°C (sublimation)
Quantity 50 g
Beilstein 3555192
Sensitivity Light sensitive
Formula Weight 204.07
Percent Purity 99%
Chemical Name or Material Trimethylsulfonium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.