missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfonium Iodide, TRC

Product Code. 30454208
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454208 1 g 1g
30453197 50 g 50g
30452222 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454208 Supplier Toronto Research Chemicals Supplier No. TRCT7974801G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2181-42-2
Molecular Formula C3 H9 S . I
Molecular Weight (g/mol) 204.07
Synonym Sulfonium, trimethyl-, iodide (1:1) (ACI), Sulfonium, trimethyl-, iodide (8CI, 9CI), Trimethylsulfonium iodide (6CI, 7CI), Trimethyl-λ[3]-sulfane hydroiodide, Trimethylsulphonium iodide
IUPAC Name trimethylsulfanium;iodide
SMILES [I-].C[S+](C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H9S.HI/c1-4(2)3;/h1-3H3;1H/q+1;/p-1
Formula Weight 203.947
Chemical Name or Material Trimethylsulfonium iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.