missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfonium tetrafluoroborate, 98+%

Product Code. 11380144
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11380144 25 g 25g
11370144 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11380144

Brand: Thermo Scientific Alfa Aesar L00647.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 676-88-0 | C3H9BF4S | 163.97 g/mol

Trimethylsulfonium tetrafluoroborate, is used as Methylene transfer agent and as a pharmaceutical intermediate. It is also used as a raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsulfonium tetrafluoroborate, is used as Methylene transfer agent and as a pharmaceutical intermediate. It is also used as a raw material in organic synthesis.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676-88-0
Molecular Formula C3H9BF4S
Molecular Weight (g/mol) 163.97
MDL Number MFCD00031582
InChI Key RNBNWTAKWJKQNN-UHFFFAOYSA-N
Synonym trimethylsulfonium tetrafluoroborate, trimethylsulphonium tetrafluoroborate 1-, trimethyl sulfonium fluoroborate, trimethylsulfanium tetrafluoroborate, trimethylsulphoniumtetrafluoroborate
PubChem CID 3083643
IUPAC Name trimethylsulfanium;tetrafluoroborate
SMILES C[S+](C)C.F[B-](F)(F)F

Specifications

Melting Point 214°C to 218°C
Quantity 25 g
UN Number UN3261
Beilstein 3708437
Solubility Information It is soluble in water.
Formula Weight 163.98
Percent Purity ≥98%
Chemical Name or Material Trimethylsulfonium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.