missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfoxonium Iodide, TRC

Product Code. 30449523
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449523 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449523 Supplier Toronto Research Chemicals Supplier No. TRCT79750010G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Trimethylsulfoxonium Iodide
CAS 1774-47-6
Molecular Formula C3H9IOS
Quantity 10 g
Synonym Trimethylsulfonium Iodide Oxide, Trimethylsulfoxonium Iodide, NSC 71213, S,S,S-Trimethylsulfoxonium iodide, Trimethyloxosulfonium Iodide, Trimethyloxosulphonium Iodide
InChI Formula InChI=1S/C3H9OS.HI/c1-5(2,3)4;/h1-3H3;1H/q+1;/p-1
SMILES [I-].C[S+](=O)(C)C
Molecular Weight (g/mol) 220.07
Formula Weight 219.9419
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.