missing translation for 'onlineSavingsMsg'
Learn More

Trimethyltin chloride, 1M solution in methylene chloride, AcroSeal™

Product Code. 10595480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10595480 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10595480 Supplier Thermo Scientific Acros Supplier No. 428711000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1066-45-1 | C3H11ClSn | 201.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1066-45-1, 75-09-2
Density 1.3560g/mL
Flash Point 113°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C3H11ClSn
MDL Number MFCD00000520
Quantity 100 mL
Specific Gravity 1.356
Synonym trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride
InChI Key FVFGWISLDLUGHX-UHFFFAOYSA-N
SMILES Cl.C[SnH](C)C
IUPAC Name chloro(trimethyl)stannane
Molecular Weight (g/mol) 201.28
PubChem CID 14016
Formula Weight 199.25
Chemical Name or Material Trimethyltin chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trimethyltin chloride, 1M solution in methylene chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.