missing translation for 'onlineSavingsMsg'
Learn More

Trimethyltin chloride, 99%

Product Code. 10488540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10488540 50 g Glass bottle 50g
10225640 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10488540 Supplier Thermo Scientific Acros Supplier No. 163990500
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1066-45-1 | C3H11ClSn | 201.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-45-1
Molecular Formula C3H11ClSn
Molecular Weight (g/mol) 201.28
MDL Number MFCD00000520
InChI Key FVFGWISLDLUGHX-UHFFFAOYSA-N
Synonym trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride
PubChem CID 14016
IUPAC Name chloro(trimethyl)stannane
SMILES Cl.C[SnH](C)C

Specifications

Melting Point 37.0°C to 39.0°C
Color White
Boiling Point 148.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SnCl
Quantity 50 g
Fieser 17,388; 03,19; 04,27; 05,26; 06,41; 07,18; 13,239
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,organic solvents
Formula Weight 199.25
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Trimethyltin chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethyltin chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.