missing translation for 'onlineSavingsMsg'
Learn More

Trimyristin, 90%

Product Code. 10599993 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10599993 5 g Glass bottle 5g
10046000 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10599993 Supplier Thermo Scientific Acros Supplier No. 422090050
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 555-45-3 | C45H86O6 | 722.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-45-3
Molecular Formula C45H86O6
Molecular Weight (g/mol) 722.59
MDL Number MFCD00036229
InChI Key DUXYWXYOBMKGIN-UHFFFAOYSA-N
Synonym trimyristin, glycerol trimyristate, myristin, propane-1,2,3-triyl tritetradecanoate, glyceryl trimyristate, dynasan 114, myristic acid triglyceride, trimyristoylglycerol, tetradecanoic acid, 1,2,3-propanetriyl ester, myristin, tri
PubChem CID 11148
ChEBI CHEBI:77391
IUPAC Name 2,3-di(tetradecanoyloxy)propyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCC

Specifications

Melting Point 54°C to 58°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 9896
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohols,benzene,chloroform,ether
Formula Weight 722.59
Percent Purity 90%
Physical Form Crystalline Powder
Chemical Name or Material Trimyristin, 90%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.