missing translation for 'onlineSavingsMsg'
Learn More

Trimyristin, 95%

Product Code. 15415129
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415129 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15415129 Supplier Thermo Scientific Alfa Aesar Supplier No. J62819.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 555-45-3 | C45H86O6 | 723.177 g/mol

It can be used for the preparation of solid lipid nanoparticles (SLN) composed of trimyristin (solid lipid) and poloxamer 407 (surfactant). It is also used as a synthesis reagent and as an emollient, refatting agent, skin conditioning agent, solvents and viscosity controlling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used for the preparation of solid lipid nanoparticles (SLN) composed of trimyristin (solid lipid) and poloxamer 407 (surfactant). It is also used as a synthesis reagent and as an emollient, refatting agent, skin conditioni
TRUSTED_SUSTAINABILITY

Specifications

CAS 555-45-3
Melting Point 56°C to 57°C
Color White
Molecular Formula C45H86O6
Linear Formula [CH3(CH2)12COOCH2]2CHOCO(CH2)12CH3
MDL Number MFCD00036229
Quantity 25 g
Beilstein 1718242
Merck Index 14,9724
Synonym trimyristin, glycerol trimyristate, myristin, propane-1,2,3-triyl tritetradecanoate, glyceryl trimyristate, dynasan 114, myristic acid triglyceride, trimyristoylglycerol, tetradecanoic acid, 1,2,3-propanetriyl ester, myristin, tri
Solubility Information Insoluble in water; Soluble in chloroform, ether, ethanol or dichloromethane
InChI Key DUXYWXYOBMKGIN-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCC
IUPAC Name 2,3-di(tetradecanoyloxy)propyl tetradecanoate
Molecular Weight (g/mol) 723.177
PubChem CID 11148
ChEBI CHEBI:77391
Formula Weight 723.16
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Trimyristin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.