missing translation for 'onlineSavingsMsg'
Learn More

Triolein, tech.

Product Code. p-7042402
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15474799

Brand: Thermo Scientific Alfa Aesar J62419.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 122-32-7 | C57H104O6 | 885.453 g/mol

Glycerol trioleate can be used as an emulsifier, also as creams, lotions and lipsticks matrix. Used as textile finishing agent in textile industry, also used as additive in pigment grind. In pharm industry, used as ointment, emulsifiable paste, suppository, lotion, emulsifier in air agent, stabilizing agent, lubricating agent, antifoaming agents. In food industry, it has the function as emulsify, disperse, defoaming, blister, antistaling of starch and control fat agglutination, usually used as agent in candy, ice cream, cookie and bread. When used in beverage, could avoid ester to float, and avoid protein to sink, therefore to improve stability. When used in bread, cookie , could improve organization structure, increase volume, antistaling, prolong warranty validity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycerol trioleate can be used as an emulsifier, also as creams, lotions and lipsticks matrix. Used as textile finishing agent in textile industry, also used as additive in pigment grind. In pharm industry, used as ointment, emulsifiable paste, suppository, lotion, emulsifier in air agent, stabilizing agent, lubricating agent, antifoaming agents. In food industry, it has the function as emulsify, disperse, defoaming, blister, antistaling of starch and control fat agglutination, usually used as agent in candy, ice cream, cookie and bread. When used in beverage, could avoid ester to float, and avoid protein to sink, therefore to improve stability. When used in bread, cookie , could improve organization structure, increase volume, antistaling, prolong warranty validity.

Solubility
Soluble in chlorofom, ether, and carbon tetrachloride

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-32-7
Molecular Formula C57H104O6
Molecular Weight (g/mol) 885.453
MDL Number MFCD00137563
InChI Key PHYFQTYBJUILEZ-BSCDBXJPSA-N
Synonym triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate
PubChem CID 45253964
IUPAC Name [2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC

Specifications

Color Yellow
Density 0.91
Boiling Point 235°C to 240°C (18 mmHg)
Flash Point 330°C (626°F)
Linear Formula (C17H33COOCH2)2CHOCOC17H33
Refractive Index 1.468
Quantity 25 g
Beilstein 1718692
Merck Index 14,9732
Solubility Information Soluble in chlorofom, ether, and carbon tetrachloride
Formula Weight 885.45
Percent Purity 75%
Grade Technical
Physical Form Liquid
Chemical Name or Material Triolein
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.