missing translation for 'onlineSavingsMsg'
Learn More

Trioxsalen, MedChemExpress

Product Code. 16478646
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478646 100 mg 100mg
16462256 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478646 Supplier Medchem Express Supplier No. HYB1157
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Trioxsalen (Trisoralen), a psoralen derivative, is a photochemical DNA crosslinker. Trioxsalen only works after photoactivation with near ultraviolet light. Trioxsalen is a photosensitizer that can be used for the research of vitiligo and hand eczema. Trioxsalen is used for visualization of genomic interstrand cross-links localized by laser photoactivation.

Trioxsalen (Trisoralen), a psoralen derivative, is a photochemical DNA crosslinker. Trioxsalen only works after photoactivation with near ultraviolet light. Trioxsalen is a photosensitizer that can be used for the research of vitiligo and hand eczema. Trioxsalen is used for visualization of genomic interstrand cross-links localized by laser photoactivation.

Specifications

Chemical Name or Material Trioxsalen
CAS 3902-71-4
Color White
Purity Grade Notes Research
Molecular Formula C14H12O3
Quantity 100 mg
Synonym Trisoralen Trioxysalen TMP
Solubility Information DMSO : 6.6 mg/mL (28.92 mM; Need ultrasonic and warming) ∣H2O : 1 mg/mL (4.38 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1C=C(C)C2=CC3=C(OC(C)=C3)C(C)=C2O1
Molecular Weight (g/mol) 228.24
Formula Weight 228.24
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.