missing translation for 'onlineSavingsMsg'
Learn More

Trioxysalen, TRC

Product Code. 30456912
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456912 100 mg 100mg
30451851 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456912 Supplier Toronto Research Chemicals Supplier No. TRCT806580100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3902-71-4
Molecular Formula C14 H12 O3
Molecular Weight (g/mol) 228.24
Synonym 7H-Furo[3,2-g][1]benzopyran-7-one, 2,5,9-trimethyl-, 5-Benzofuranacrylic acid, 6-hydroxy-β,2,7-trimethyl-, δ-lactone (6CI,7CI), 2,5,9-Trimethyl-7H-furo[3,2-g][1]benzopyran-7-one, 4,2',8-Trimethylpsoralen, 4,5',8-Trimethylpsoralen, 4,5',8-Trimethylpsoralene, NSC 71047, TMP, Trimethylpsoralen, Trioxsalen, Trioxysalen, Trisoralen
IUPAC Name 2,5,9-trimethylfuro[3,2-g]chromen-7-one
SMILES CC1=CC(=O)Oc2c(C)c3oc(C)cc3cc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H12O3/c1-7-4-12(15)17-14-9(3)13-10(6-11(7)14)5-8(2)16-13/h4-6H,1-3H3
Formula Weight 228.0786
Chemical Name or Material Trioxsalen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.