missing translation for 'onlineSavingsMsg'
Learn More

Tripalmitin, 99%

Product Code. 10608074 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10608074 1 g Glass bottle 1g
10368300 5 g Glass bottle 5g
10082970 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10608074 Supplier Thermo Scientific Acros Supplier No. 215670010
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 555-44-2 | C51H98O6 | 807.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-44-2
Molecular Formula C51H98O6
Molecular Weight (g/mol) 807.33
MDL Number MFCD00008995
InChI Key PVNIQBQSYATKKL-UHFFFAOYSA-N
Synonym tripalmitin, glycerol tripalmitate, tripalmitoylglycerol, glyceryl tripalmitate, palmitic triglyceride, tripalmitate, triglyceryl palmitate, barolub, dynasan 116, propane-1,2,3-triyl tripalmitate
PubChem CID 11147
ChEBI CHEBI:77393
IUPAC Name 2,3-di(hexadecanoyloxy)propyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 65°C to 68°C
Color White
Boiling Point 315°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula [CH3(CH2)14CO2CH2]2CHO2C(CH2)14CH3
Quantity 1 g
Beilstein 02, IV, 1176
Merck Index 15, 9908
Solubility Information Solubility in water: insoluble. Other solubilities: very soluble in ether,benzene,chloroform,soluble in toluene
Formula Weight 807.33
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tripalmitin, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Tripalmitin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.