missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenyl-d15 Phosphate, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Triphenylphosphate D15 |
| CAS | 1173020-30-8 |
| Molecular Formula | C18 2H15 O4 P |
| Quantity | 0.5 g |
| Synonym | Phen-2,3,4,5,6-d5-ol, 1,1',1''-phosphate, Triphenylphosphate D15 |
| InChI Formula | InChI=1 S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D,13 D,14 D,15 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(OP(=O)(Oc2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H])Oc3c([2 H])c([2 H])c([2 H])c([2 H])c3[2 H])c([2 H])c1[2 H] |
| IUPAC Name | tris(2,3,4,5,6-pentadeuteriophenyl) phosphate |
| Molecular Weight (g/mol) | 341.38 |
| Formula Weight | 341.1649 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction